Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ovalbumin Rabbit anti-Chicken, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB1225100UL
Description
Detect Ovalbumin using this Anti-Ovalbumin Antibody, Egg white validated for use in ELISA, western blot, immunohistochemistry.
Specifications
Ovalbumin | |
Polyclonal | |
Immunoglobulin purified by delipidation, salt fractionation and ion-exchange chromatography. Liquid in 0.02M Potassium Phosphate buffer, 0.15M Sodium Chloride, pH 7.2, with 0.01% sodium azide. | |
Ovalbumin (Hen Egg White) | |
Primary | |
−20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
1 mg | |
Chicken | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction