Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-oxoacid CoA transferase 2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89008048 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-048 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-048 Supplier Abnova Corporation Supplier No. H00064064B01

Mouse polyclonal antibody raised against a full-length human OXCT2 protein.

OXCT2 is a testis-specific succinyl-CoA:3-oxoacid CoA transferase (EC 2.8.3.5), which catalyzes the reversible transfer of CoA from succinyl-CoA to acetoacetate in the first step of ketone body utilization. See also OXCT1 (MIM 601424).[supplied by OMIM

Sequence: MAALRLLASVLGRGVPAGGSGLALSQGCARCFATSPRLRAKFYADPVEMVKDISDGATVMIGGFGLCGIPENLIAALLRTRVKDLQVVSSNVGVEDFGLGLLLAARQVRRIVCSYVGENTLCESQYLAGELELELTPQGTLAERIRAGGAGVPAFYTPTGYGTLVQEGGAPIRYTPDGHLALMSQPREVREFNGDHFLLERAIRADFALVKGWKADRAGNVVFRRSARNFNVPMCKAADVTAVEVEEIVEVGAFPPEDIHVPNIYVDRVIKGQKYEKRIERLTILKEEDGDAGKEEDARTRIIRRAALEFEDGMYANLGIGIPLLASNFISPSMTVHLHSENGILGLGPFPTEDEVDADLINAGKQTVTVLPGGCFFASDDSFAMIRGGHIQLTMLGAMQVSKYGDLANWMIPGKKVKGMGGAMDLVSSQKTRVVVTMQHCTKDNTPKIMEKCTMPLTGKRCVDRIITEKAVFDVHRKKELTLRELWEGLTVDDIKKSTGCAFAVSPNLRPMQ

Specifications

Antigen 3-oxoacid CoA transferase 2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human OXCT2 protein.
Formulation No additive
Gene OXCT2
Gene Accession No. BC030015.1
Gene Alias FKSG25/FLJ00030/SCOT-T
Gene Symbols OXCT2
Host Species Mouse
Immunogen OXCT2 (AAH30015.1, 1 a.a. ∼ 513 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Endocrine/Metabolism
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 64064
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.