Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oxytocin Rabbit anti-Human, Hamster, Mouse, Rat, Rabbit, Sheep, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB911
Description
Anti-Oxytocin Antibody is an antibody against Oxytocin for use in DB, immunocytochemistry, immunohistochemistry (paraffin), RIA.
Specifications
Oxytocin | |
Polyclonal | |
Neat rabbit antisera. Liquid. Contains no preservative. | |
Rabbit | |
Unpurified | |
Primary | |
Hamster, Human, Mouse, Rabbit, Rat, Sheep | |
Serum |
Dot Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA) | |
Unconjugated | |
OXT; MGC126890; MGC126892; OT; OT-NPI | |
Synthetic oxytocin (Sigma) conjugated to thyroglobulin. | |
100 μL | |
NM_000915.2 | |
−20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction