Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P-Cadherin Mouse anti-Human, Clone: 6A9, MilliporeSigma™

Catalog No. 05916MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-916-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-916-MI Supplier MilliporeSigma Supplier No. 05916
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-P-Cadherin Antibody, clone 6A9 is validated for use in immunoprecipitation, western blot, IC for the detection of P-Cadherin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P-Cadherin
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 6A9
Conjugate Unconjugated
Gene Accession No. NM_001793
Gene Symbols CDH3, P-cadherin, Placental-cadherin, HJMD, CDHP, PCAD
Host Species Mouse
Immunogen P-Cadherin extracellular domain purified from A431 cell membranes.
Purification Method Ascites
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for 24 months.
Form Ascites
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.