Learn More
Human P-Cadherin PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Supplier:  R&D Systems FAB861P
Catalog No. FAB861P
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
P-Cadherin Monoclonal specifically detects P-Cadherin in Human samples. It is validated for Flow Cytometry, Knockout Validated.
Specifications
Specifications
P-Cadherin
Monoclonal
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CAD3, cadherin 3, P-cadherin (placental), cadherin 3, type 1, P-cadherin (placental), cadherin-3, CDH3, CDHP, CDHPcalcium-dependent adhesion protein, placental, HJMD, PCAD, PCadherin, PCADP-cadherin, Placental cadherin
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
1001
Human
Purified
Flow Cytometry, KnockDown
104805
Flow Cytometry 10 uL/10^6 cells, Knockout Validated 10 uL/10^6 cells
CAA45177
CDH3
Mouse myeloma cell line NS0-derived recombinant human P-Cadherin Asp108-Gly654 Accession # CAA45177
100 Tests
Primary
Detects human P-Cadherin in direct ELISAs and Western blots. In Western blots, does not cross-react with recombinant human (rh) Cadherin-8, recombinant mouse P-Cadherin, or rhVE-Cadherin.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.