Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P-glycoprotein Mouse anti-Human, PE, Clone: 17F9, BD

Catalog No. BDB557003 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557003 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557003 Supplier BD Biosciences Supplier No. 557003
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with a 170-180kDa transmembrane glycoprotein (P-glycoprotein), a product of the multidrug resistance-1 (MDR1) gene. This glycoprotein is expressed on MDR positive cells and has been reported to be expressed on many normal tissues, such as adrenal glands and endothelium, in the brain and skin. P-glycoprotein is known to impart drug resistance to cells by pumping many anti-cancer drugs out of the cytoplasm. 17F9 antibody binds to the surface of the viable, unfixed MDR+ cell line, PMG-Y (please inquire), and is able to partially block the binding of UIC2 antibody (another MDR-specific monoclonal antibody). Immunoprecipitation application is reported, but not routinely tested in house.

Host Species: Mouse
Clone: 17F9
Isotype: IgG2b, κ
Cross Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen P-glycoprotein
Applications Flow Cytometry
Classification Monoclonal
Clone 17F9
Conjugate PE
Description CD243; ABCB1; ABC20; CLCS; GP170; MDR1; P-GP; PGY1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human MDR1 Transfected Cell Line
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.