Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P-Selectin Mouse anti-Human, Clone: P8G6, MilliporeSigma™

Catalog No. MAB2154MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2154MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2154MI Supplier MilliporeSigma Supplier No. MAB2154
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect P-Selectin using this Anti-P-Selectin Antibody, clone P8G6 validated for use in ELISA, immunoprecipitation, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P-Selectin
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone P8G6
Conjugate Unconjugated
Formulation Purified in 0.02M PBS, 0.25M NaCl, pH 7.6 with 0.1% NaN3. Liquid
Gene Accession No. NM_003006.3
Gene Symbols SELPLG, CLA, PSGL-1, CD162, PSGL1
Host Species Mouse
Immunogen IL-1beta activated HUVEC (human)
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.