Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P-Selectin Glycoprotein Ligand-1 Mouse anti-Human, Clone: KPL-1, MilliporeSigma™

Catalog No. MAB4092MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4092MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4092MI Supplier MilliporeSigma Supplier No. MAB4092
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect P-Selectin Glycoprotein Ligand-1 using this Anti-P-Selectin Glycoprotein Ligand-1 Antibody, clone KPL-1 validated for use in flow cytometry, immunohistochemistry, immunohistochemistry (paraffin), immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P-Selectin Glycoprotein Ligand-1
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone KPL-1
Conjugate Unconjugated
Formulation Purified Immunoglobulin. Liquid in 0.02M PB with 0.25M NaCl, pH7.6, 0.1% Sodium Azide.
Gene Accession No. NM_003006.3
Gene Symbols SELPLG, CLA, PSGL-1, CD162, PSGL1
Host Species Mouse
Immunogen The antibody epitope was mapped to a site within a consensus tyrosine sulfation motif of PSGL-1, previously shown to be essential for interaction with P-selectin (and now shown to be essential for recognition of PSGL-1 by L-selectin).
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.