Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p115 Mouse, Unlabeled, Clone: 15, BD

Catalog No. BDB612260 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612260 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612260 Supplier BD Biosciences Supplier No. 612260
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Maturation and post translational modification of proteins occurs after their biosynthesis at the endoplasmic reticulum and their transport through the Golgi apparatus. The process involves the transport of vesicles carrying the proteins through a vectorial process of vesicle budding and fusion from the cis-compartment to the medial-compartment and the trans-compartment of the Golgi apparatus. p115 is a 959 amino acid protein located at the Golgi apparatus that, with the NEM-sensitive fusion protein and the soluble NSF attachment protein (SNAP), is required for vesicle transport from the cis-compartment to the medial-compartment. p115 protein is related to the yeast Uso1p essential for the vesicular transport from the endoplasmic reticulum to the Golgi. Native p115 appears to be a homo-oligomer, with two globular heads and a tail that resemble the overall structure of myosin. p115 is extracted from the Golgi apparatus with high salt or high pH, indicative of a membrane associated protein. p115 interacts with the golgi matrix protein GM130 but this interaction is disrupted by the Golgi fragmentation during mitosis and the phosphorylation of GM130.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen p115
Applications Western Blot
Classification Monoclonal
Clone 15
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat p115 aa. 843-955
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.