Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p120 Catenin Mouse, Unlabeled, Clone: 98, BD

Catalog No. BDB610133 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610133 50 μg
BDB610134 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610133 Supplier BD Biosciences Supplier No. 610133
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The membrane associated protein p120 Catenin (pp120 (Src substrate); p120cas) was identified as a tyrosine kinase substrate that is phosphorylated in Src transformed cells or in response to growth factor stimulation. It shares structural similarity with the Drosophila Armadillo protein and the vertebrate β-catenin and γ-catenin proteins. This similarity is evidenced by its characteristic Arm domain that is composed of 42-amino acid motif repeats. In the cell, p120 Catenin is localized to the E-Cadherin/catenins cell adhesion complex. Like β- and γ-catenin, p120 Catenin directly associates with the cytoplasmic C-terminus of E-Cadherin via its Arm domain and may similarly interact with other Cadherins. It exists as four isoforms that range in size from 90-115kDa. Expression of these isoforms is heterogeneous in human carcinomas, suggesting that altered pp120 expression contributes to malignancy due to loss of functional cell adhesions. Multiple tyrosine residues (Y96, Y112, Y228, Y280, Y257, Y291, Y296, and Y302) in p120 Catenin are phosphorylated by Src and these phosphorylations may facilitate interaction with PTP1C/SHP-1 in response to EGF stimulation. Thus, p120 Catenin is an Arm domain protein that interacts with both cell adhesion molecules, such as cadherins and cell signaling molecules, such as PTP1C.

Host Species: Mouse
Clone: 98
Isotype: IgG1
Species Reactivity: Human
Immunogen: Mouse pp120 aa. 326-632
Formula Weight [Chemical]: 120kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen p120 Catenin
Applications Western Blot
Classification Monoclonal
Clone 98
Concentration 250μg/mL
Conjugate Unconjugated
Description pp120 (Src Substrate); p120cas
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse pp120 aa. 326-632
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.