Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p120 Catenin (pT916) Mouse anti-Human, PE, Clone: 1/CATENIN, BD

Catalog No. BDB558637 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558637 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558637 Supplier BD Biosciences Supplier No. 558637

Mouse Monoclonal Antibody

The membrane associated protein pp120 Src substrate (p120 catenin, p120cas) was identified as a tyrosine kinase substrate that is phosphorylated in Src-transformed cells. It shares structural similarity with the Drosophila Armadillo protein and the vertebrate β-catenin and γ-catenin proteins in its 42-amino acid Arm domain. p120 catenin is localized to the E-Cadherin/catenin cell adhesion complex. Like βand γ-catenin, p120 catenin directly associates with the cytoplasmic C-terminus of E-Cadherin via its Arm domain. It exists as four isoforms that range in size from 90 to 115kDa. Expression of these isoforms is heterogeneous in human carcinomas, suggesting that altered expression contributes to malignancy. Phosphorylation of multiple serine (S252, S268, S288, and S879), and threonine (T310 and T916) residues in p120 catenin may regulate its activity. The S879 residue is phosphorylated after PKC activation, while the S268 site is dephosphorylated after PKC activation. The latter residue is phosphorylated in vitro by p160 Rock. S252 and T310 residues are phosphorylated in vitro by GSK3b. Thus, p120 catenin function may be regulated in a complex manner through both serine and threonine phosphorylation. The 1/Catenin monoclonal antibody recognizes the phosphorylated T916 in the carboxy-terminal tail of human p120 catenin. The orthologous

Intracellular Staining, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen p120 Catenin (pT916)
Applications Flow Cytometry
Classification Monoclonal
Clone 1/CATENIN
Conjugate PE
Host Species Mouse
Immunogen Phosphorylated Human p120 Catenin Peptide
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.