Learn More
p120 Catenin (pY228) Mouse, Unlabeled, Clone: 21A, BD
Mouse Monoclonal Antibody
$373.50
Specifications
Antigen | p120 Catenin (pY228) |
---|---|
Clone | 21A |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
The membrane associated protein pp120 Src substrate (p120 Catenin, p120cas)was identified as a tyrosine kinase substrate that is phosphorylated in Src transformed cells or in response to growth factor stimulation. It shares structural similarity with the Drosophila Armadillo protein and the vertebrate β-catenin and γ-catenin proteins. This similarity is evidenced by its characteristic Arm domain that is composed of 42-amino acid motif repeats. In the cell, p120 Catenin is localized to the E-Cadherin/catenins cell adhesion complex. Like β- and γ-catenin, p120 Catenin directly associates with the cytoplasmic C-terminus of E-Cadherin via its Arm domain and may similarly interact with other Cadherins. It exists as four isoforms that range in size from 90-115kDa. Expression of these isoforms is heterogeneous in human carcinomas, suggesting that altered pp120 expression contributes to malignancy due to loss of functional cell adhesions. Multiple tyrosine residues (Y96, Y112, Y228, Y280, Y257, Y291, Y296, and Y302) in p120 Catenin are phosphorylated by Src and these phosphorylations may facilitate interaction with PTP1C/SHP-1 in response to EGF stimulation. Thus, p120 Catenin is an Arm domain protein that interacts with both cell adhesion molecules, such as cadherins and cell signaling molecules, such as PTP1C.
Flow Cytometry, Immunofluorescence, Intracellular Staining, Western Blotting
Specifications
p120 Catenin (pY228) | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
21A | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
Mouse p120 Catenin (pY228) | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.