Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p130 [Cas] Mouse, Unlabeled, Clone: 21, BD

Catalog No. BDB610272 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610272 150 μg
BDB610271 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610272 Supplier BD Biosciences Supplier No. 610272
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p47 [v-crk] is the product of a transforming gene, v- crk , that was isolated from avian sarcoma viruses. The v-crk protein is a fusion product of the viral gag protein and a part of cellular crk that includes SH2 and SH3 domains. v-crk induced transformation increases tyrosine phosphorylation of several cellular proteins, including p130 [Cas]. p130 [Cas] is tightly associated with v-crk via the SH2 domain of v-crk. Tyrosine phosphorylation of p130 [Cas] occurs in conjunction with cellular transformation in cells that express v-src or v-crk. This phosphorylation leads to a change in p130 [Cas] localization from the cytoplasm to the cell membrane and, possibly, to the nucleus. Since p130 [Cas] also associates with v-src, it may be a v-src substrate.

TRUSTED_SUSTAINABILITY

Specifications

Antigen p130 [Cas]
Applications Western Blot
Classification Monoclonal
Clone 21
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.