Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p130Cas (pY249) Mouse anti-Human, Unlabeled, Clone: J169-757.12.2, BD

Catalog No. BDB558401 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558401 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558401 Supplier BD Biosciences Supplier No. 558401
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p47v-crk (v-Crk) is the product of a transforming gene, v-crk, that was isolated from avian sarcoma viruses. The v-Crk protein is a fusion product of viral Gag protein and a part of cellular Crk that includes SH2 and SH3 domains. v-Crk-induced transformation increases tyrosine phosphorylation of several cellular proteins, including p130Cas (CRK-associated substrate). The p130Cas is tightly associated with v-Crk via the SH2 domain of v-Crk. Tyrosine phosphorylation of p130Cas occurs in conjunction with cellular transformation in cells that express v-Src or v-Crk. This phosphorylation leads to a change in p130Cas localization from the cytoplasm to the cell membrane and, possibly, to the nucleus. Since p130Cas also associates with v-Src, it may be a v-Src substrate. Several phosphorylation sites have been described in p130Cas upon Fibroblast Growth Factor stimulation, and phosphorylated tyrosine (Y249) might function as a binding site for the Crk-adaptor molecule. The J169-757.12.2 monoclonal antibody recognizes the phosphorylated Y249 of human p130Cas. The orthologous phosphorylation sites in mouse and rat p130Cas are Y253 and Y347, respectively.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen p130Cas (pY249)
Applications Western Blot
Classification Monoclonal
Clone J169-757.12.2
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human p130Cas
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.