Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-p14ARF (Ab-1) (118-132), Polyclonal
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ PC409100UL
Description
Specifically detects p14ARF (118-132) Clone: in Human samples, and it is validated for Immuno Blotting
The human p14ARF protein (murine homologue is p19ARF) results from the translation of an alternative reading frame of the CDKN2A (p16INK4) gene. Theα transcript gives rise to the p16 gene product while theβ transcript gives rise to the p14ARF gene product. Although transcriptionally derived from shared exons, there is little amino acid sequence homology between the two proteins. The function of the p14ARF protein is to activate a p53 response and thereby induce cell cycle arrest. p53 in turn negatively regulates p14ARF expression. The mechanism of action for p14ARF is a direct binding of the COOH terminal domain of MDM2 which in turn forms a ternary complex with p53. Interaction with MDM2 results in the blocking of the MDM2 induced degradation of p53. p14ARF transcription is also modulated by the activity of E2F1 expression. Loss of p14ARF correlates with loss of p16 expression and alteration in expression of both proteins is frequently seen in non-small cell lung carcinoma.
Specifications
p14ARF (118-132) | |
Polyclonal | |
Undiluted serum. | |
a synthetic peptide [(K)AGRARCLGPSARGPG] corresponding to amino acids 118-132 of human p14ARF (p16β) | |
RUO | |
Recognizes the ∽14kDa p14ARF protein in Saos-2 cells. Sold under license of U.S. Patent 6,482,929. | |
−20°C | |
IgG |
Immunoblot | |
Unconjugated | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction