Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p150 [Glued] Mouse, Unlabeled, Clone: 1, BD

Catalog No. BDB610473 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610473 50 μg
BDB610474 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610473 Supplier BD Biosciences Supplier No. 610473
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p150 [Glued] was identified as a polypeptide associated with cytoplasmic dynein, the minus-end-directed microtubule-based motor protein. p150 [Glued] is also a member of the oligomeric dynactin complex. Dynactin mediates dynein-driven vesicle motility, as well as lower eukaryote nuclear transport. p150 [Glued] bears significant homology to the product of the Glued gene in Drosophila. It has been shown in vitro to be a required activator of dynein-mediated transport along microtubules. The p150 [Glued] component of the dynactin complex binds to microtubules and the actin-like protein Centractin (Arp-1), another member of the dynactin complex. In the developing rat, p150 [Glued] is expressed at high levels in neural tissue. Microtubule bindings assays with selected constructs of p150 [Glued] indicate that amino acids 39-150 are required for microtubule association.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen p150 [Glued]
Applications Western Blot
Classification Monoclonal
Clone 1
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.