Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p14ARF/CDKN2A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | p14ARF/CDKN2A |
---|---|
Concentration | 2.5 mg/mL |
Dilution | Western Blot 2 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11059085T
![]() |
Novus Biologicals
NB11059085SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB11059085
![]() |
Novus Biologicals
NB11059085 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
p14ARF/CDKN2A Polyclonal specifically detects p14ARF/CDKN2A in Human samples. It is validated for Western Blot.Specifications
p14ARF/CDKN2A | |
Western Blot 2 ug/ml | |
Polyclonal | |
Rabbit | |
Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Transcription Factors and Regulators, Tumor Suppressors | |
Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
ARF, CDK4 inhibitor p16-INK4, CDK4IP14ARF, CDKN2, cell cycle negative regulator beta, CMM2P16-INK4A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A, cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4), INK4, INK4A, MLMP16INK4, MTS-1, MTS1P14, Multiple tumor suppressor 1, p14, p14ARF, P16, p16-INK4, p16-INK4a, P16INK4A, P19, p19Arf, TP16 | |
CDKN2A | |
IgG | |
Affinity Purified | |
16 kDa |
2.5 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
Q8N726 | |
1029 | |
A synthetic peptide made to a portion of human p14ARF (between residues 100-173). [Swiss-Prot# Q8N726] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title