Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p190 Mouse, Unlabeled, Clone: 30, BD

Catalog No. BDB610149 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610149 50 μg
BDB610150 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610149 Supplier BD Biosciences Supplier No. 610149
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GTPase activating proteins (GAPs) stimulate the GTP-hydrolyzing activity of GTPases, such as p21[ ras]. Ras-GAP (120kDa) is perhaps the best characterized GAP. p190 is a Ras-GAP associated protein that is tyrosine phosphorylated in transformed and growth factor-stimulated cells. Ras-GAP and p190 are targets of oncoproteins and growth factor receptors. Recombinant p190 enhances GTPase activity of the rho family of GTPases. The complete sequence of p190 exhibits three major regions: an N-terminal region with homology to all GTPases, a middle region homologous to transcriptional repressors, and a C-terminal region the is similar to n-chimerin and other small GTPases.

TRUSTED_SUSTAINABILITY

Specifications

Antigen p190
Applications Western Blot
Classification Monoclonal
Clone 30
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Frog, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.