Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p21-Arc Mouse, Unlabeled, Clone: 26, BD

Catalog No. BDB612234 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612234 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612234 Supplier BD Biosciences Supplier No. 612234
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cellular morphology, adhesion, and motility occur through the reorganization of the actin cytoskeleton. This reorganization of actin filaments occurs through the interactions between actin and actin binding proteins. Actin-binding proteins regulate the polymerization and depolymerization of actin, connect actin-based structures to membranes and to other cytoskeletal elements, power the movement of actin filaments, and cross-link actin filaments into bundles. Actin related proteins (Arp) 2/3 complex is an actin polymerization inducing complex that includes Arp2, Arp3, p41-Arc, p34-Arc, p21-Arc, p20 Arc, and p16-Arc. The Arp2 and Arp3 subunits may nucleate actin polymerization, while the p41-Arc subunit is a WD repeat-containing protein that may regulate both the activity and localization of the Arp2/3 complex. Arp3, p34-Arc, and p21-Arc are localized to the lamellipodia of stationary and locomoting fibroblasts. Both WASP and Abp1p are acidic sequence-containing proteins that activate the Arp2/3 complex. However, WASP binds actin monomers, while the endocytosis-related Abp1p protein binds actin filaments. Thus, Arp2/3 complex may regulate actin polymerization in specific cell locations through interaction with actin binding Arp2/3 activators.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen p21-Arc
Applications Western Blot
Classification Monoclonal
Clone 26
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human p21-Arc aa. 10-118
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.