Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human p21/CIP1/CDKN1A Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1047
This item is not returnable.
View return policy
Description
p21/CIP1/CDKN1A Monoclonal specifically detects p21/CIP1/CDKN1A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Intracellular Staining by Flow Cytometry.Specifications
p21/CIP1/CDKN1A | |
Monoclonal | |
Unconjugated | |
Supplied as a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CAP20cyclin-dependent kinase inhibitor 1, CDK-interacting protein 1, CDKN1A, CDKN1melanoma differentiation associated protein 6, CIP1, CIP1WAF1CDK-interaction protein 1, cyclin-dependent kinase inhibitor 1A (p21, Cip1), MDA6, MDA-6, Melanoma differentiation-associated protein 6, p21, p21CIP1, p21Cip1/Waf1, PIC1, SDI1DNA synthesis inhibitor, wild-type p53-activated fragment 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1026 | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunoprecipitation, Flow Cytometry | |
195720 | |
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 4 ug/500 ug cell lysate, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells | |
P38936 | |
CDKN1A | |
E. coli-derived recombinant human p21/CIP1/CDKN1A Ser2-Pro164 Accession # P38936 | |
200 μg | |
Primary | |
Detects human p21/CIP1/CDKN1A in direct ELISAs and Western blots. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction