Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p21/WAF1/Cip1 Mouse anti-Human, Clone: CP74, MilliporeSigma™

Catalog No. 05655MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-655-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-655-MI Supplier MilliporeSigma Supplier No. 05655
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-p21/WAF1/Cip1 Antibody, clone CP74 is a high quality Mouse Monoclonal Antibody for the detection of p21/WAF1/Cip1 and has been validated in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p21/WAF1/Cip1
Applications Western Blot
Classification Monoclonal
Clone CP74
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Gene Accession No. NM_000389
Gene Symbols CDKN1A, MDA-6, SDI1, MDA6, p21, p21CIP1, PIC1, CIP1, P21, CDKN1, WAF1, CAP20
Host Species Mouse
Immunogen Recombinant full-length human Cip1 with an N-terminal histidine tag; epitope corresponds to amino acids 1-80 of full-length human Cip1
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.