Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-p21WAF1 Clone: DF10,

Catalog No. OP68100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
OP68100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OP68100UG Supplier MilliporeSigma™ Supplier No. OP68100UG

Mouse Monoclonal Antibody

Specifically detects p21WAF1 Clone: DF10 in Human samples, and it is validated for Immunoprecipitation

The tumor suppressor p53 transcriptionally activates a number of genes including the WAF1/CIP1 gene in response to DNA damage. The ∽21kDa product of the WAF1 gene is found in a complex involving cyclins, cyclin dependent kinases (CDK), and PCNA in normal cells but not transformed cells and appears to be a universal inhibitor of CDK activity. One consequence of p21WAF1 binding to and inhibiting CDKs is to prevent CDK-dependent phosphorylation and subsequent inactivation of the Rb protein which is essential for cell cycle progression. p21WAF1 is, therefore, a potent and reversible inhibitor of cell cycle progression at both the G1 and G2 checkpoints, presumably to allow sufficient time for DNA repair to be completed. Irreversible G1 or G2 arrest leads to apoptosis. However, the role of p21WAF1 in apoptosis is less clear although p53-mediated apoptosis leads to increased WAF1 expression. Induction of p21WAF1 in response to DNA damage can occur by both p53-dependent and p53-independent mechanisms, in response to mitogenic stimuli, differentiation, or in tumor cells with mutated p53. Functional p21WAF1 is essential for p53-mediated G1 arrest presumably due to WAF1 inhibition of both CDK activity and PCNA-dependent DNA replication. WAF1 has also been identified as a gene involved in cellular senescence, termed sdi1. Not surprisingly, p21WAF1 overexpression is growth suppressive, consistent with its role as an inhibitor of CDKs. By inhibiting Rb inactivation in a p53-dependent fashion, p21WAF1 serves to integrate cell cycle control mediated by p53 and Rb.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p21WAF1
Applications Immunoprecipitation
Classification Monoclonal
Clone DF10
Conjugate Unconjugated
Formulation In 0.05M sodium phosphate buffer, 0.2% gelatin.
Host Species Mouse
Immunogen full-length, recombinant human p21WAF1
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽21kDa p21WAF1 protein in skin or colon tissue and in cells expressing wild-type p53 (e.g. Hs27 or U205 cells treated with DNA damaging agents).
Target Species Human
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.