Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p21WAF1 Mouse anti-Human, Clone: EA10, MilliporeSigma™

Catalog No. MABE325MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE325MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE325MI Supplier MilliporeSigma Supplier No. MABE325
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-p21WAF1 Antibody, clone EA10 is a Mouse Monoclonal Antibody for detection of p21WAF1 and has been validated in western blot, flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p21WAF1
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone EA10
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ supernatant in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_000380
Gene Symbols CDKN1A, CAP20, CDKN1, CIP1, MDA6, PIC1, SDI1, WAF1
Host Species Mouse
Immunogen Recombinant protein corresponding to human p21WAF1.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.