Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p21waf1 Mouse anti-Bovine, Human, Rat, Clone: HZ52, MilliporeSigma™

Catalog No. MAB88058MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB88058MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB88058MI Supplier MilliporeSigma Supplier No. MAB88058

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Use Anti-p21waf1 Antibody, clone HZ52 (Mouse Monoclonal Antibody) validated in flow cytometry, immunoprecipitation, immunohistochemistry (paraffin) to detect p21waf1 also known as Cip1, MDA-6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p21waf1
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone HZ52
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid containing 10mM PBS, pH 7.4, with 0.2% BSA and 0.09% sodium azide.
Gene Accession No. NM_000389.2; NM_078467.1
Gene Symbols CDKN1A, P21, MDA-6, SDI1, MDA6, p21, p21CIP1, PIC1, CIP1, CDKN1, WAF1, CAP20
Host Species Mouse
Immunogen Full length human recombinant p21 protein.
Quantity 50 μg
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.