Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-p23/PTGES3, Clone: JJ6, R&D Systems™

Catalog No. A450
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A450 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A450 Supplier R&D Systems Supplier No. A450
This item is not returnable. View return policy

Applications:

Western Blotting

Specifications

Antigen p23/PTGES3
Applications Western Blot
Classification Monoclonal
Clone JJ6
Conjugate Unconjugated
Dilution Western Blot
Formulation Supplied as a solution in PBS containing Glycerol and Sodium Azide. with Sodium Azide
Gene Alias CPGES, cPGESp23, Cytosolic prostaglandin E2 synthase, Hsp90 co-chaperone, P23cytosolic prostaglandin E synthase, Progesterone receptor complex p23, prostaglandin E synthase 3, prostaglandin E synthase 3 (cytosolic), PTGES3, TEBP, TEBPEC 5.3.99.3, Telomerase-binding protein p23, unactive progesterone receptor, 23 kD
Gene Symbols PTGES3
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human p23/PTGES3
Purification Method Protein A or G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10728
Test Specificity Detects human p23/PTGES3 in Western blot.
Target Species Human
Content And Storage Store the unopened product at -20 to -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.