Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p230 trans Golgi Mouse anti-Human, Unlabeled, Clone: 15, BD

Catalog No. BDB611280 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611280 50 μg
BDB611281 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611280 Supplier BD Biosciences Supplier No. 611280
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Golgi apparatus is a very complex and dynamic organelle that functions in protein sorting and modification. Numerous structural and regulatory proteins are involved in the budding, docking, and fusion of Golgi-directed vesicles. p230 trans Golgi is a peripheral membrane protein associated with the cytosolic face of the trans Golgi network (TGN). Protein sequence analysis indicates that it is highly hydrophilic and contains two proline-rich regions, as well as a high frequency of heptad repeats which are characteristic of α-helices that form dimeric coiled-coiled structures. Additionally, p230 trans Golgi contains a ESLALEELEL sequence, a motif of the acidic granin proteins that are found in secretory granules of neuroendocrine cells. p230 trans Golgi cycles between the cytosol and the TGN via non-clathrin coated vesicles in a G protein-dependent manner. The localization of p230 trans Golgi to the TGN is dependent on aromatic residues in its C-terminal non-coiled coil domain. Thus, p230 trans-Golgi is aTGN-specific protein that is important for the biogenesis of distinct populations of non-clathrin coated vesicles and, thus, functions in vesicular transport from the TGN.

Host Species: Mouse
Clone: 15
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human p230 trans Golgi aa. 2063-2179
Formula Weight [Chemical]: 230kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen p230 trans Golgi
Applications Western Blot
Classification Monoclonal
Clone 15
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human p230 trans Golgi aa. 2063-2179
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.