Learn More
Description
Kip1 is a cyclin-dependent kinase (cdk) inhibitor that was identified as a result of its role in TGFβ-induced G1 phase arrest and cell-cell contact. In vitro , p27 [Kip1] binds tightly to Cyclin D-Cdk4, Cyclin E-Cdk2, and Cyclin A-Cdk2 complexes and inhibits their activity. In normal cells, Kip1 is sequestered and its activity gradually decreases as cells reach S phase. The addition of TGF-β in early G1 blocks this decline in activity, by preventing the sequestration of Kip1. TGF-β also reduces the levels of Cdk4. Kip1 preferentially binds Cyclin D-Cdk4, but the lower levels of Cdk4 in TGF-β treated cells allow Kip1 to be available for binding to Cyclin E-Cdk2 and Cyclin A-Cdk2. Kip1 is structurally related to Cip1/WAF1, having a similar 60 amino acid sequence in the N-terminal region. A 52 amino acid Kip1 peptide (residues 28-79) from this region is sufficient to inhibit cdk activity in vitro.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
Specifications
| Antigen | p27[Kip1] |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 57 |
| Concentration | 250μg/mL |
| Conjugate | Unconjugated |
| Formulation | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.