Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

purinergic receptor P2X, ligand-gated ion channel, 5, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017921 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-921 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-921 Supplier Abnova Corporation Supplier No. H00005026D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human P2RX5 protein.

The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel. Several characteristic motifs of ATP-gated channels are present in its primary structure, but, unlike other members of the purinoceptors family, this receptor has only a single transmembrane domain. Three transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq

Sequence: MGQAGCKGLCLSLFDYKTEKYVIAKNKKVGLLYRLLQASILAYLVVWVFLIKKGYQDVDTSLQSAVITKVKGVAFTNTSDLGQRIWDVADYVIPAQGENVFFVVTNLIVTPNQRQNVCAENEGIPDGACSKDSDCHAGEAVTAGNGVKTGRCLRRENLARGTCEIFAWCPLETSSRPEEPFLKEAEDFTIFIKNHIRFPKFNFSKSNVMDVKDRSFLKSCHFGPKNHYCPIFRLGSVIRWAGSDFQDIALEGGVIGINIEWNCDLDKAASECHPHYSFSRLDNKLSKSVSSGYNFRFARYYRDAAGVEFRTLMKAYGIRFDVMVNGKGAFFCDLVLIYLIKKREFYRDKKYEEVRGLEDSSQEAEDEASGLGLSEQLTSGPGLLGMPEQQELQEPPEAKRGSSSQKGNGSVCPQLLEPHRST

Specifications

Antigen purinergic receptor P2X, ligand-gated ion channel, 5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human P2RX5 protein.
Formulation PBS with no preservative; pH 7.4
Gene P2RX5
Gene Accession No. NM_002561.2
Gene Alias MGC47755/P2X5/P2X5R
Gene Symbols P2RX5
Host Species Rabbit
Immunogen P2RX5 (NP_002552.2, 1 a.a. ∼ 422 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5026
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.