Learn More
Description
Specifications
Specifications
| Antigen | P2X3/P2RX3 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen 1:100-1:1000 |
| Formulation | Whole antisera with 0.05% Sodium Azide |
| Gene Alias | ATP receptor, MGC129956, P2X purinoceptor 3, P2X receptor, subunit 3, P2X3purinergic receptor P2X3, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 3, purinoceptor P2X3 |
| Gene Symbols | P2RX3 |
| Host Species | Guinea Pig |
| Immunogen | Synthetic peptide: VEKQSTDSGAYSIGH, corresponding to amino acids 383-397 of Rat P2X3. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
