Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2X3 Receptor Rabbit anti-Rat, Primate, Human, Polyclonal, MilliporeSigma™

Catalog No. AB5895MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5895MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5895MI Supplier MilliporeSigma Supplier No. AB5895

Rabbit Polyclonal Antibody

Detect P2X3 Receptor using this Anti-P2X3 Receptor Antibody, pain validated for use in immunocytochemistry, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen P2X3 Receptor
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit serum. Liquid. Contains 0.05% sodium azide.
Gene Symbols P2RX3; MGC129956; P2X3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminus of rat P2X3.
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002559.2
Target Species Human, Primate, Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.