Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2Y1 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB9263
Description
Anti-P2Y1 Antibody is an antibody against P2Y1 for use in immunohistochemistry (paraffin).
Specifications
P2Y1 | |
Polyclonal | |
Affinity purified immunoglobulin. Liquid in PBS, pH 7.7 with 0.01% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
P2RY1; P2Y1 | |
Synthetic peptide from the C-terminal domain of human P2Y1. | |
50 μg | |
NM_002563.2 | |
−70°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction