Learn More
Description
Specifications
Specifications
| Antigen | P2Y13 Receptor |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.05% sodium azide as a preservative. Reconstitute with 200μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min). |
| Gene Symbols | P2RY13; P2Y13; FKSG77; SP174; GPR94; GPCR1; GPR86 |
| Host Species | Rabbit |
| Immunogen | Purified peptide from amino acids 119-134 of human P2Y13 |
| Purification Method | Affinity Purified |
| Quantity | 200 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.