Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-p300 Clone: NM-11,

Catalog No. NA46100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
NA46100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NA46100UG Supplier MilliporeSigma™ Supplier No. NA46100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects p300 Clone: NM-11 in Human, Rat samples, and it is validated for Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Immuno Blotting

p300 is a nuclear phosphoprotein which was originally shown to specifically bind to adenovirus E1A protein. The binding of p300 to E1A occurs in the conserved region 1 (CR1) of E1A at a site which is close to but non-overlapping with that for the Rb family of proteins including p105 and p107. p300 has been shown to bind to the extreme N-terminal region requiring the presence of positively charged residues at positions 2 and 3 of E1A via a zinc-finger motif in the C-terminal portion of p300. While the binding of p300 and Rb to E1A is independent, it has been shown that upon binding, E1A mediates the physical contact between the two proteins. The p300 protein is actively phosphorylated in both quiescent and proliferating cells however the amount of phosphorylation increases during progression from late G1 intoM phase of the cell cycle. In addition to the demonstrated binding of p300 to E1A it has also been shown to bind to other molecules including DNA (in a site specific manner) and to components of the TATA binding protein complexes (TBP). The TBP complex association is most likely with TFIID and occurs in a protein-protein dependent rather than a DNA dependent manner and, in such a complex, p300 functions as a co-activator of transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p300
Applications Immunoblot, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone NM-11
Conjugate Unconjugated
Formulation In 0.05M sodium phosphate buffer, 50% glycerol.
Host Species Mouse
Immunogen recombinant, human p300 protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the p300 protein in A431, HeLa, and ML-1 cells.
Target Species Human, Rat
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.