Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p31comet Mouse anti-Human, Clone: E29.19.14, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABE451
Description
Detect p31comet using this mouse monoclonal antibody, Anti-p31comet Antibody, clone E29.19.14 validated for use in western blotting and immunoprecipitation.
Specifications
p31comet | |
Monoclonal | |
Unconjugated | |
NP_001003690 | |
Mouse | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunoprecipitation, Western Blot | |
E29.19.14 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
MAD2L1BP, CMT2 | |
Recombinant protein corresponding to human p31comet. | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction