Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p32 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB2991MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2991MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2991MI Supplier MilliporeSigma Supplier No. AB2991
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-p32 Antibody is validated for use in western blot, IC for the detection of p32.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p32
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Symbols C1QBP; C1qBP; GC1QBP; HABP1; SF2P32; SF2p32; gC1Q-R; gC1qR; p32; p33
Host Species Rabbit
Immunogen Cocktail of peptides corresponding to the N-terminus of human and mouse p32.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_001203
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.