Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p38α Mouse, Unlabeled, Clone: 27, BD

Catalog No. BDB612168 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612168 50 μg
BDB612169 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612168 Supplier BD Biosciences Supplier No. 612168

Mouse Monoclonal Antibody

Activation of the immune and inflammatory responses often involves the recognition of bacterial endotoxin (lipopolysaccharide or LPS). Binding of LPS by monocytic cells results in the production and release of proinflammatory cytokines, such as IL-1 and TNF-α. LPS-induced signaling cascades involve members of the Ser/Thr protein kinase family known as the mitogen activated protein kinases (MAPKs). One of these, p38 MAP kinase (p38 MAPK or CSPB2), is a human homologue of HOG1, a yeast MAP kinase that is essential for growth under conditions of elevated osmolarity. Similar to HOG1, p38 MAPK is phosphorylated in response to hyperosmolarity, but also in response to a variety of stimuli including LPS, IL-1, or TNF-α. Efficient activation of p38 MAPK requires phosphorylation of Thr-180 and Tyr-182. At least three Thr/Tyr kinases (MKK3, MKK4/SEK1, and MKK6) phosphorylate and activate p38 MAPK. This leads to the activation of multiple transcription factors (NF-κB, ATF-2, Elk-1, and CHOP/GADD143) that induce expression of proinflammatory cytokine genes. In addition to its role in immunity, p38 MAPK is widely expressed and serves as an important signal transducer in a variety of cell types.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen p38α
Applications Western Blot
Classification Monoclonal
Clone 27
Concentration 250μg/mL
Conjugate Unconjugated
Description SAPK2a
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human p38 MAPK aa. 243-355
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.