Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-p38 MAP Kinase (341-360), Polyclonal

Catalog No. 506123200UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
50-612-3200UL 200 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-612-3200UL Supplier MilliporeSigma™ Supplier No. 506123200UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects p38 MAP Kinase (341-360) Clone: in Human, Mouse, Rat samples, and it is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Immuno Blotting

p38 MAP kinase (MAPK), also called RK and CSBP, is the mammalian homologue of the yeast HOG kinase and participates in a cascade controlling cellular responses to cytokines and stress. Like the SAPK/JNK pathway, p38 MAPK is activated by a variety of cellular stresses including osmotic shock, inflammatory cytokines, lipopolysaccharides (LPS), UV light and growth factors. Both MKK3 and SEK phosphorylate p38 on tyrosine and threonine at the sequence TGY resulting in p38 activation. Activated p38 MAPK has been shown to phosphorylate and activate MAPKAP kinase-2 and to phosphorylate the transcription factors ATF-2 and Max.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p38 MAP Kinase (341-360)
Applications Flow Cytometry, Immunoblot, Immunoblot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation In 150mM NaCl, 10mM HEPES, 50% glycerol, 0.01% BSA, pH 7.5.
Host Species Rabbit
Immunogen a synthetic peptide (TYDEYISFVPPPLDQEEMES) corresponding to amino acids 341-360 of human p38 MAP kinase
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽38kDa p38 MAPK protein. Antibody Target Gene Symbol: MAPK14 Target Synonym: CRK1, CSBP, CSBP1, CSBP2, CSPB1, EXIP, Hog, MAPK p38, MGC102436, MGC105413, MXI2, P38, P38 KINASE, P38 Map Kinase, p38 Mapk alpha, P38-ALPHA, p38-RK, p38/Hog1, p38/Mpk2, P38/RK, p38a, p38Hog, p38MAPK, PRKM14, PRKM15, RK, SAPK2A Entrez Gene Name: mitogen-activated protein kinase 14
Target Species Human, Mouse, Rat
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.