Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p38 alpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP219662
Description
p38 alpha Polyclonal antibody specifically detects p38 alpha in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay.Specifications
| p38 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 1% BSA, 20% Glycerol (pH7) with 0.025% Proclin 300 | |
| CSAID-binding protein, Csaids binding protein, CSBP, CSBP1MAX-interacting protein 2, CSBP2, CSPB1, cytokine suppressive inflammatory drug binding protein, Cytokine suppressive inflammatory drug-binding protein, EC 2.7.11, EC 2.7.11.24, EXIP, MAP kinase 14, MAP kinase MXI2, MAP kinase p38 alpha, MAPK 14, mitogen-activated protein kinase 14, Mitogen-activated protein kinase p38 alpha, Mxi2, p38, p38 MAP kinase, p38 mitogen activated protein kinase, p38ALPHA, p38alpha Exip, PRKM14, PRKM15, RK, SAPK2A, stress-activated protein kinase 2A | |
| Rabbit | |
| 41 kDa | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Immunology, Innate Immunity, MAP Kinase Signaling, Protein Kinase, Signal Transduction | |
| 1432 | |
| Canine, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.64mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Knockout Validated | |
| Q16539 | |
| MAPK14 | |
| Full length human p38 MAPK Recombinant protein. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity:Bovine (100%), Chimpanzee (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction