Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p40-phox Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. 07503MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-503-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-503-MI Supplier MilliporeSigma Supplier No. 07503

Rabbit Polyclonal Antibody

Anti-p40-phox Antibody is an antibody against p40-phox for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p40-phox
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Protein A purified antibody in 0.1M Tris-Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH 7.4.
Gene Symbols NCF4; SH3PXD4; p40-phox; OTTHUMP00000043756; MGC3810; NCF; P40PHOX; p40phox; OTTHUMP00000028736; NCF-4
Host Species Rabbit
Immunogen C-terminal 6HIS-tagged fusion protein corresponding to full length human p40-phox
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_013416
Target Species Human, Rat
Content And Storage 2-8°C for 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.