Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p47[phox] Mouse anti-Human, Unlabeled, Clone: 1, BD

Catalog No. BDB610354 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610354 50 μg
BDB610355 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610354 Supplier BD Biosciences Supplier No. 610354
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The neutrophil respiratory burst oxidase (NADPH-oxidase) generates superoxide and secondary oxygen-derived toxic products in response to bacteria or a variety of soluble stimuli. The enzyme is dormant in resting neutrophils. The active site of this enzyme is located in an integral membrane cytochrome, b558, which consists of the two subunits gp91[phox] and p21[phox]. Superoxide production depends on the formation of a complex that includes two cytosolic proteins, p67[phox] and p47[phox]. The GTP-binding protein Rac is also an essential component for oxidase activity. p47[phox] is a highly basic protein that contains two SH3 domains. The C-terminal quarter of the molecule contains many potential phosphorylation sites, consisting of serines and basic residues. Expression of p47[phox] is restricted to cells of phagocytic or lymphocytic lineage. IFN-γ is a potent inducer of both p47[phox] mRNA and protein. p47[phox] is an early reactant in oxidase assembly and this assembly can be inhibited by a C-terminal peptide of the large subunit of cytochrome b558. It is thought that p47[phox] binds directly to the cytochrome, while p67[phox] associates with the cytochrome by binding p47[phox].

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen p47[phox]
Applications Western Blot
Classification Monoclonal
Clone 1
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.