Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-p53 Binding Protein 1 (Ab-1), Polyclonal

Catalog No. PC712100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
PC712100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PC712100UL Supplier MilliporeSigma™ Supplier No. PC712100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects p53 Binding Protein 1 Clone: in Human, Mouse samples, and it is validated for Immunofluorescence, Immunoprecipitation

53BP1 is a protein that binds to the DNA-binding domain of p53 and enhances p53-mediated transcriptional activation. It has been reported that 53BP1 is an ATM substrate involved in early DNA damage-signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p53 Binding Protein 1
Applications Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation In 100mM NaCl, 50mM Tris-HCl, 20% glycerol, pH 7.6.
Host Species Rabbit
Immunogen a recombinant protein consisting of human 53BP1 fused to a His·Tag™ sequence
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽230kDa 53BP1 protein in HeLa cell nuclear extract
Target Species Human, Mouse
Content And Storage ≤ -70°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.