Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p53 Mouse anti-Human, Unlabeled, Clone: DO-7, BD

Catalog No. BDB554294 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554294 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554294 Supplier BD Biosciences Supplier No. 554294
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p53 is a 53kDa nuclear phosphoprotein that acts as a tumor suppressor protein, and is involved in inhibiting cell proliferation when DNA damage occurs. The gene for p53 is the most commonly mutated gene yet identified in human cancers. Missense mutations occur in tumors of the colon, lung, breast, ovary, bladder and several other organs. The mutant p53 is overexpressed in a variety of transformed cells and wildtype p53 forms specific complexes with several viral oncogenes including SV40 large T, E1B from adenovirus, and E6 from human papilloma virus. Wildtype p53 plays a role as a checkpoint protein for DNA damage during the G1/S-phase of the cell cycle. However, it is still unclear, whether point mutated forms of p53 are simple null mutants and/or dominant negatively acting proteins. The DO-7 antibody recognizes human wildtype and mutant p53. It cross-reacts with bovine p53 but does not cross-react with mouse or rat p53. DO-7 recognizes an epitope between amino acids 1-45 of known forms of human p53. Human recombinant p53 protein was used as immunogen. Wildtype 53 proteins have a very short half-life and are usually not detectable with monoclonal antibodies (mAbs) in normal tissues.

Host Species: Mouse
Clone: DO-7
Isotype: IgG2b
Species Reactivity: Human
Immunogen: Human Recombinant p53 protein
Formula Weight [Chemical]: 53kDa

Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen p53
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone DO-7
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Recombinant p53 protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Bovine, Human, Monkey
Content And Storage Store undiluted at 4°C
Isotype IgG2b
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.