Learn More
Description
The gene for the nuclear phosphoprotein p53 is the most commonly mutated gene yet identified in human cancers. Missense mutations occur in tumors of the colon, lung, breast, ovary, bladder and several other organs. The mutant p53 is over-expressed in a variety of transformed cells and it forms specific complexes with several viral oncogenes including SV40 large T, E1B from adenovirus and E6 from human papilloma virus. Recent data suggest that wild type p53 plays a role as a checkpoint protein for DNA damage during the S-phase of the cell cycle. However, it is still unclear whether point mutated forms of p53 are simple null mutants and/or dominant negatively acting proteins. p53 migrates at a reduced molecular weight of 53kDa. Clone PAb 1801 recognizes an epitope between amino acids 32-79 in the N-terminal domain of human wild type and mutant p53 antibody. It does not cross-react with p53 from other species. A truncated recombinant human p53 fusion protein was used as immunogen.
Host Species: Mouse
Clone: PAb 1801
Isotype: IgG1
Species Reactivity: Human
Immunogen: Recombinant fusion protein
Formula Weight [Chemical]: 53kDa
Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting
Specifications
Specifications
| Antigen | p53 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | PAB 1801 |
| Concentration | 0.5mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant fusion protein |
| Purification Method | Affinity Purified |
| Quantity | 0.25 mg |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.