Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p53 Mouse anti-Bovine, Chicken, Human, Hamster, Mouse, Monkey, Rat, Clone: PAb240, MilliporeSigma™

Catalog No. CBL404MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL404MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL404MI Supplier MilliporeSigma Supplier No. CBL404
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect p53 with Anti-p53 Antibody, aa 211-220, clone240 (Mouse Monoclonal Antibody), that has been shown to work in immunoprecipitation, western blotting and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p53
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone PAb240
Conjugate Unconjugated
Formulation Purified mouse monoclonal in buffer containing 0.1M Tris-glycine (pH 7.4), 0.15M NaCl, with 0.05% sodium azide. We recommend that each laboratory determine an optimum working titre for use in its particular application.
Gene Accession No. NP_000537
Gene Symbols TP53, TRP53, P53, p53, LFS1
Host Species Mouse
Immunogen Murine p53-beta galactosidase fusion protein expressed in E. coli.
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Chicken, Hamster, Human, Monkey, Mouse, Rat
Content And Storage 4°C for up to 1 month, −20°C in undiluted aliquots for long term
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.