Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p53 with control Mouse, PE, Clone: G59-12, BD

Catalog No. BDB557027 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557027 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557027 Supplier BD Biosciences Supplier No. 557027
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p53 is a 53 kD nuclear phosphoprotein that acts as a tumor suppressor protein, and is involved in inhibiting cell proliferation when DNA damage occurs. The gene for p53 is the most commonly mutated gene yet identified in human cancers. Missense mutations occur in tumors of the colon, lung, breast, ovary, bladder and several other organs. The mutant p53 is overexpressed in a variety of transformed cells and wild-type p53 forms specific complexes with several viral oncogenes including SV40 large T, E1B from adenovirus, and E6 from human papilloma virus. Wild type p53 plays a role as a checkpoint protein for DNA damage during the G1/S-phase of the cell cycle.

G59-12 recognizes mutant and wild type human, rat and mouse p53 tumor suppressor protein. Recombinant full-length human p53 was used as immunogen. The G59-12 clone was originally characterized by western blot analysis, immunoprecipitation and immunohistochemical staining. Clone MOPC-21 is a mouse IgG1 isotypecontrol. The MOPC-21 antibody has unknown specificity.

TRUSTED_SUSTAINABILITY

Specifications

Antigen p53 with control
Applications Flow Cytometry
Classification Monoclonal
Clone G59-12
Conjugate PE
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Includes PE Mouse Anti-p53 (100 tests), PE Mouse IgG1, κ Isotype Control (100 tests)
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.