Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p53 Upregulated Modulator of Apoptosis (PUMA) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABC158MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC158MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC158MI Supplier MilliporeSigma Supplier No. ABC158

Rabbit Polyclonal Antibody

Anti-p53 Upregulated Modulator of Apoptosis (PUMA) Antibody detects level of p53 Upregulated Modulator of Apoptosis (PUMA) and has been published and validated for use in Western Blotting and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p53 Upregulated Modulator of Apoptosis (PUMA)
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols BBC3; PUMA
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the N-terminus of human p53 Upregulated Modulator of Apoptosis (PUMA).
Purification Method Affinity Purified
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_055232
Target Species Human
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.