Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p54nrb/NonO Mouse anti-Human, Rat, Clone: 78-1-C6, MilliporeSigma™

Catalog No. 05950MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-950-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-950-MI Supplier MilliporeSigma Supplier No. 05950
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-p54nrb/NonO Antibody, clone 78-1-C6 is a high quality Mouse Monoclonal Antibody for the detection of p54nrb/NonO and has been validated in immunoprecipitation, western blotting and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p54nrb/NonO
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 78-1-C6
Conjugate Unconjugated
Gene Accession No. NM_007363
Gene Symbols NONO, NRB54, nrb, NMT55, p54nrb, p54, P54, NRB, P54NRB
Host Species Mouse
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids TVREAGPPAFYRPNS
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.