Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p57[Kip2] Mouse anti-Human, Unlabeled, Clone: A120-1, BD

Catalog No. BDB556346 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556346 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556346 Supplier BD Biosciences Supplier No. 556346
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cyclin dependent kinase inhibitors (CdkIs) inhibit progression through the cell cycle by binding to cyclins, Cdks, or cyclin-Cdk complexes. CdkIs are classified into two groups based on protein structure. p57[Kip2] belongs to a group that also includes p27[Kip1] and p21 (also known as Sdi1, Cip1, Waf1 and Pic1). Members of this group have a homologous amino-terminal Cdk inhibitory domain. Members of the second group [p16 (INK4A), p15 (INK4B), p18 (INK4C), and p19 (INK4D)] contain ankyrin repeat motifs. p57[Kip2] is a potent, tight-binding inhibitor of several G1 and S phase cyclin-Cdk complexes including cyclin E-Cdk2, cyclin D2-Cdk4, and cyclin A-Cdk2. It inhibits the mitotic cyclin B1-Cdk1 complex to a lesser extent. mRNA studies suggest that p57[Kip2] expression is tissue-specific, the highest levels have been found in embryonic and adult skeletal muscle, heart, kidney, lung, eye and brain. This is in contrast to the widespread tissue expression of p27[Kip1] and p21 mRNA. The expression of p57[Kip2] to primarily terminally differentiated cells suggests that p57[Kip2] may play a specialized role in cell cycle control. Clone A120-1 reacts with human p57[Kip2]. Recombinant human p57[Kip2] was used as immunogen.

Host Species: Mouse
Clone: A120-1
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: p57[Kip2] Recombinant Human

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen p57[Kip2]
Applications Western Blot
Classification Monoclonal
Clone A120-1
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen p57[Kip2] Recombinant Human
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.