Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p67 (phox) Mouse, Unlabeled, Clone: 29, BD

Catalog No. BDB610912 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610912 50 μg
BDB610913 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610912 Supplier BD Biosciences Supplier No. 610912
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The neutrophil respiratory burst oxidase (NADPH-oxidase) generates superoxide and secondary oxygen-derived toxic products in response to bacteria or a variety of soluble stimuli. The active site of this enzyme is located in an integral membrane cytochrome, b558, that consists of the two subunits gp91 [phox] and p21 [phox]. Superoxide production depends on the formation of a complex that includes p67 [phox], p47 [phox], and the GTP-binding protein Rac. Upon activation, these proteins translocate from the cytosol to the membrane where they assemble with b558 and induce oxidase activity. p67 [phox] contains two SH3 domains and binds, via its C-terminal SH3 domain, to the proline rich
region of p47 [phox]. This binding allows p67 [phox] to indirectly associate with the oxidase. It is thought that the phosphorylated forms of p67 [phox] and p47 [phox] interact and that the phosphorylation of p67 [phox] is regulated by both PKC-dependent and independent pathways. Although the role of p67 [phox] in electron flow control is poorly understood, it is thought that it regulates the transfer of electrons from NADPH to reduce flavin.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen p67 (phox)
Applications Western Blot
Classification Monoclonal
Clone 29
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human p67 [phox] aa. 317-469
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at -20°C.
Isotype IgG2b
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.