Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-p73 Ab 2, Clone: ER-15,

Catalog No. OP109100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
OP109100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OP109100UG Supplier MilliporeSigma™ Supplier No. OP109100UG

Mouse Monoclonal Antibody

Specifically detects p73 Clone: ER-15 in Human, Monkey samples, and it is validated for Gel Supershift, Immunoprecipitation, Immuno Blotting

The p73 protein shares considerable structural and functional similarity to p53. The p73 gene encodes 4 polypeptides through alternative splicing of exons 11-13 with p73α and p73β expressed in all tissues while expression of p73γ and p73δ may be more restricted. All isoforms show significant sequence identity to the transactivation, DNA binding, and oligomerization domains of p53, and many of the critical amino acid residues in p53 essential for DNA binding are conserved in p73. Not surprisingly, p73 binds to a p53 consensus sequence and transactivates many, but not all, p53-responsive genes. Overproduction of p73 leads to growth arrest and induction of apoptosis in a p53-independent manner. However, p73 does not transactivate all p53-responsive genes and the level of transactivation is often lower than that by p53; thus, the mechanisms of growth arrest and induction of apoptosis by p73 are at least partially distinct from p53. p73 does not appear to be targeted by many of the viral oncoproteins such as E1B 55K, SV40 T antigen, and HPV E6 that bind to and inactivate p53, however both p53 and p73 do bind the Adenovirus E4orf6 protein, suggesting that p73 may be independently inactivated by viral oncoproteins. All p73 isoforms bind one another and p73α and p73β appear to bind p53 as well. Interestingly, mutant p53 binds p73α and inhibits transcriptional activation by p73 indicating that mutational inactivation of p53 may serve to simultaneously inactivate p73. One key difference between p53 and p73 is that, unlike p53, p73 does not appear to be activated in response to DNA damage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p73
Applications Gel Supershift, Immunoblot, Immunoblot, Immunoprecipitation
Classification Monoclonal
Clone ER-15
Conjugate Unconjugated
Formulation In 0.05M sodium phosphate buffer, 0.2% gelatin.
Host Species Mouse
Immunogen recombinant, human p73α protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽80kDa α and the ∽68kDa β isoforms of p73 in HEK293, LOS, and IM9 cells. Will immunoprecipitate p73α/p73β complexes.
Target Species Human, Monkey
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.